Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nudix (nucleoside diphosphate linked moiety X)-type motif 12, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008288 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-288 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-288 Supplier Abnova Corporation Supplier No. H00083594B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human NUDT12 protein.

Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances (Abdelraheim et al., 2003 [PubMed 12790796]).[supplied by OMIM

Sequence: MSSVKRSLKQEIVTQFHCSAAEGDIAKLTGILSHSPSLLNETSENGWTALMYAARNGHPEIVQFLLEKGCDRSIVNKSRQTALDIAVFWGYKHIANLLATAKGGKKPWFLTNEVEECENYFSKTLLDRKSEKRNNSDWLLAKESHPATVFILFSDLNPLVTLGGNKESFQQPEVRLCQLNYTDIKDYLAQPEKITLIFLGVELEIKDKLLNYAGEVPREEEDGLVAWFALGIDPIAAEEFKQRHENCYFLHPPMPALLQLKEKEAGVVAQARSVLAWHSRYKFCPTCGNATKIEEGGYKRLCLKEDCPSLNGVHNTSYPRVDPVVIMQVIHPDGTKCLLGRQKRFPPGMFTCLAGFIEPGETIEDAVRREVEEESGVKVGHVQYVACQPWPMPSSLMIGCLALAVSTEIKVDKNEIEDARWFTREQVLDVLTKGKQQAFFVPPSRAIAHQLIKHWIRINPNL

Specifications

Antigen nudix (nucleoside diphosphate linked moiety X)-type motif 12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human NUDT12 protein.
Formulation No additive
Gene NUDT12
Gene Accession No. NM_031438.2
Gene Alias DKFZp761I172
Gene Symbols NUDT12
Host Species Mouse
Immunogen NUDT12 (NP_113626.1, 1 a.a. ∼ 462 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Endocrine/Metabolism
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 83594
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.