Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT2, Mouse, Clone: 4A4-3C3, Abnova™

Catalog No. 89016159 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-159 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-159 Supplier Abnova Corporation Supplier No. H00000318M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant NUDT2.

This gene encodes a member of the MutT family of nucleotide pyrophosphatases, a subset of the larger NUDIX hydrolase family. The gene product possesses a modification of the MutT sequence motif found in certain nucleotide pyrophosphatases. The enzyme asymmetrically hydrolyzes Ap4A to yield AMP and ATP and is responsible for maintaining the intracellular level of the dinucleotide Ap4A, the function of which has yet to be established. This gene may be a candidate tumor suppressor gene. Alternative splicing has been observed at this locus and three transcript variants, all encoding the same protein, have been identified. [provided by RefSeq

Sequence: MALRACGLIIFRRCLIPKVDNNAIEFLLLQASDGIHHWTPPKGHVEPGEDDLETALRETQEEAGIEAGQLTIIEGFKRELNYVARNKPKTVIYWLAEVKDYDVEIRLSHEHQAYRWLGLEEACQLAQFKEMKAALQEGHQFLCSIEA

Specifications

Antigen NUDT2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4A4-3C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant NUDT2.
Formulation PBS with no preservative; pH 7.4
Gene NUDT2
Gene Accession No. BC004926
Gene Alias APAH1/MGC10404
Gene Symbols NUDT2
Host Species Mouse
Immunogen NUDT2 (AAH04926, 1 a.a. ∼ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 318
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.