Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nudix (nucleoside diphosphate linked moiety X)-type motif 21, Mouse, Clone: 2G4-6F11, Abnova™

Catalog No. 89001201 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-201 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-201 Supplier Abnova Corporation Supplier No. H00011051M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant NUDT21.

The protein encoded by this gene is one subunit of a cleavage factor required for 3' RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3' end processing complex and facilitates the recruitment of other processing factors. This gene encodes the 25kD subunit of the protein complex, which is composed of four polypeptides. [provided by RefSeq]

Sequence: MSVVPPNRSQTGWPRGVTQFGNKYIQQTKPLTLERTINLYPLTNYTFGTKEPLYEKDSSVAARFQRMREEFDKIGMRRTVEGVLIVHEHRLPHVLLLQLGTTFFKLPGGELNPGEDEVEGLKRLMTEILGRQDGVLQDWVIDDCIGNWWRPNFEPPQYPYIPAHITKPKEHKKLFLVQLQEKALFAVPKNYKLVAAPLFELYDNAPGYGPIISSLPQLLSRFNFIYN

Specifications

Antigen nudix (nucleoside diphosphate linked moiety X)-type motif 21
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G4-6F11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant NUDT21.
Formulation PBS with no preservative; pH 7.4
Gene NUDT21
Gene Accession No. BC001403
Gene Alias CFIM25/CPSF5/DKFZp686H1588
Gene Symbols NUDT21
Host Species Mouse
Immunogen NUDT21 (AAH01403, 1 a.a. ∼ 227 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 11051
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.