Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NuMA Mouse, Unlabeled, Clone: 22, BD

Catalog No. BDB610562 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610562 150 μg
BDB610561 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610562 Supplier BD Biosciences Supplier No. 610562
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NuMA (Nuclear Mitotic Apparatus protein) is a 2115 amino acid protein with a coiled-coil structure similar to that of myosins and intermediate filaments. Indirect immunofluorescence assays indicate that NuMA's localization is very dynamic. During interphase, NuMA is in the nucleus and during mitosis it moves to the polar regions of the mitotic spindle. NuMA is a very abundant phosphoprotein and antibodies to this protein are often found in patients with autoimmune diseases. Although NuMA is thought to be a structural component of the nucleus, the precise cellular function for this protein is still unknown.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NuMA
Applications Western Blot
Classification Monoclonal
Clone 22
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgM
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.