Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nuclear mitotic apparatus protein 1, Mouse, Clone: 1C5, Abnova™

Catalog No. 89014157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-157 Supplier Abnova Corporation Supplier No. H00004926M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NUMA1.

Sequence: SPASPMGDILQTPQFQMRRLKKQLADERSNRDELELELAENRKLLTEKDAQIAMMQQRIDRLALLNEKQAASPLEPKELEELRDKNESLTMRLHETLKQCQDLKTEK

Specifications

Antigen nuclear mitotic apparatus protein 1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NUMA1.
Formulation PBS with no preservative; pH 7.4
Gene NUMA1
Gene Accession No. NM_006185
Gene Alias NUMA
Gene Symbols NUMA1
Host Species Mouse
Immunogen NUMA1 (NP_006176, 200 a.a. ∼ 306 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 4926
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.