Learn More
Description
Specifications
Specifications
| Antigen | NuMA1 |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide. |
| Gene Symbols | NUMA1; NUMA |
| Host Species | Rabbit |
| Immunogen | Three different peptides corresponding to multiple locations of NuMA1 isoform 1. Peptide, region between amino acids 1-240. Peptide 2, amino acids 1300-1500. Peptide 3, amino acids 1860-2115. Note that these peptides are also found in isoform 2. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.