Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nup88 Mouse anti-Human, Unlabeled, Clone: 22, BD

Catalog No. BDB611896 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611896 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611896 Supplier BD Biosciences Supplier No. 611896
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nucleocytoplasmic transport involves the binding and interaction of several cytosolic and nuclear pore proteins by at least two different mechanisms. One mechanism is mediated by karyopherins to transport proteins with the nuclear localization sequence (NLS) and an alternative pathway uses the protein Transportin. Morphologically both pathways require the use of the nuclear pore complex (NPC) which acts as a gate mediating active transport of proteins and RNA into and out of the nucleus. The NLS binds to karyopherin α, and to the N-terminal region of karyopherin β. Both karyopherins bind to repeat sequences of nucleoporins at the nuclear envelope. p62 is the best characterized member of a group of nucleoporins that line the central region of the NPC. A tightly associated complex is formed by p62 and two other nucleoporins, p54 and p58. Nup88 is a nonglycosylated nucleoporin, that does not contain the characteristic GLFG or XFXFG repeats found in other NPC proteins. The structure of Nup88 includes a coiled-coil domain required for association with the NPC. Nup88 is overexpressed during oncogenesis and development, and forms a complex with the nuclear transport factors, Exportin-1 and Nup214.

Host Species: Mouse
Clone: 22
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human Nup88 aa. 314-425

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nup88
Applications Western Blot
Classification Monoclonal
Clone 22
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Nup88 aa. 314-425
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.