Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nucleoporin 98kDa, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005879 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-879 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-879 Supplier Abnova Corporation Supplier No. H00004928B02
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human NUP98 protein.

Signal-mediated nuclear import and export proceed through the nuclear pore complex (NPC), which is comprised of approximately 50 unique proteins collectively known as nucleoporins. The 98kDa nucleoporin is generated through a biogenesis pathway that involves synthesis and proteolytic cleavage of a 186kDa precursor protein. This cleavage results in the 98kDa nucleoporin as well as a 96kDa nucleoporin, both of which are localized to the nucleoplasmic side of the NPC. Rat studies show that the 98kDa nucleoporin functions as one of several docking site nucleoporins of transport substrates. The human gene has been shown to fuse to several genes following chromsome translocatons in acute myelogenous leukemia (AML) and T-cell acute lymphocytic leukemia (T-ALL). This gene is one of several genes located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in several transcript variants; however, not all variants have been fully described. [provided by RefSeq

Sequence: MKLYQTPLELKLKHSTVHVDELCPLIVPNLGVAVIHDYADWVKEASGDLPEAQIVKHWSLTWTLCEALWGHLKELDSQLNEPREYIQILERRRAFSRWLSCTATPQIEEEVSLTQKNSPVEAVFSYLTGKRISEACSLAQQSGDHRLALLLSQFVGSQSVRELLTMQLVDWHQLQADSFIQDERLRIFALLAGKPVWQLSEKKQINVCSQLDWKRSLAIHLWYLLPPTASISRALSMYEEAFQNTSDSDRYACSPLPSYLEGSGCVIAEEQNSQTPLRDVCFHLLKLYSDRHYDLNQLLEPRSITADPLDYRLSWHLWEVLRALNYTHLSAQCEGVLQASYAGQLESEGLWEWAIFVLLHIDNSGIREKAVRELLTRHCQLLETPESWAKETFLTQKLRVPAKWIHEAKAVRAHMESDKHLEALCLFKAEHWNRCHKLIIRHLASDAIINENYDYLKGFLEDLAPPERSSLIQDWETSGLVYLDYIRVIEMLRHIQQVDCSGNDLEQLHIKVTSLCSRIEQIQCYSAKDRLAQSDMAKRVANLLRVVLSLHHPPDRTSDSTPDPQRVPLRLLAPHIGRLPMPEDYAMDELRSLTQSYLRELAVGSL

Specifications

Antigen nucleoporin 98kDa
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human NUP98 protein.
Formulation No additive
Gene NUP98
Gene Accession No. BC012906.1
Gene Alias ADIR2/NUP196/NUP96
Gene Symbols NUP98
Host Species Mouse
Immunogen NUP98 (AAH12906.1, 1 a.a. ∼ 606 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4928
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.