Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nucleoporin like 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016987 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-987 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-987 Supplier Abnova Corporation Supplier No. H00009818B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human NUPL1 protein.

This gene encodes a member of the nucleoporin family that shares 87% sequence identity with rat nucleoporin p58. The protein is localized to the nuclear rim and is a component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Sequence: MSTGFSFGSGTLGSTTVAAGGTSTGGVFSFGTGASSNPSVGLNFGNLGSTSTPATTSAPSSGFGTGLFGSKPATGFTLGGTNTGIATTITTGLTLGTPATTSAATTGFSLGFNKPAASATPFALPITSTSASGLTLSSALTSTPAASTGFTLNNLGGTTATTTTASTGLSLGGALAGLGGSLFQSTNTGTSGLGQNALGLTLGTTAATSTAGNEGLGGIDFSSSSDKKSDKTGTRPEDSKALKDENLPPVICQDVENLQKFVKEQKQVQEEISRMSSKAMLKVQEDIKALKQLLSLAANGIQRNTLNIDKLKIETAQELKNAEIALRTQKTPPGLQHEYAAPADYFRILVQQFEVQLQQYRQQIEELENHLATQANNSHITPQDLSMAMQKIYQTFVALAAQLQSIHENVKVLKEQYLGYRKMFLGDAVDVFETRRAEAKKWQNTPRVTTGPTPFSTMPNAAAVAMAATLTQQQQPATGPQPSLGVSFGTPFGSGIGTGLQSSGLGSSNLGGFGTSSGFGCSTTGASTFGFGTTNKPSGSLSAGFGSSSTSGFNFSNPGITASAGLTFGVSNPASAGFGTGGQLLQLKKPPAGNKRGKR

Specifications

Antigen nucleoporin like 1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human NUPL1 protein.
Formulation PBS with no preservative; pH 7.4
Gene NUPL1
Gene Accession No. NM_014089.3
Gene Alias KIAA0410/PRO2463
Gene Symbols NUPL1
Host Species Mouse
Immunogen NUPL1 (NP_054808.1, 1 a.a. ∼ 599 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9818
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.