Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nur77 Mouse anti-Mouse, Unlabeled, Clone: 12.14, BD

Catalog No. BDB554088 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554088 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554088 Supplier BD Biosciences Supplier No. 554088
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nur77 (also known as NGF1-B, N10, TISI) is a growth factor-inducible orphan member of the steroid/thyroid hormone receptor superfamily. This superfamily encodes ligand-dependent transcription factors with a centrally located, highly conserved DNA-binding domain containing two zinc-fingers. Although Nur77 binds no known ligand, it is constitutively active when synthesized. Nur77 was originally identified as an immediate-early gene rapidly activated by serum stimulation of quiescent fibroblasts. It has since been shown to be activated by diverse signals including membrane depolarization, nerve growth factor, chemically induced seizures, adrenocorticotrophic hormone (ACTH), pentylene tetrazole, forskolin and cAMP. Nur77, like other immediate-early genes such as c-myc, has also been shown to have a role in apoptosis. Apoptosis is an internal, programmed cell death which takes place during normal development. Nur77 has been demonstrated to be required for in vitro T-cell-receptor (TCR) mediated negative selection. Negative selection, or the clonal deletion of thymocytes, normally occurs by apoptosis following engagement of the TCR. Nur77 is present in high levels in T-cell hybrids and thymocytes undergoing apoptosis, but not in growing T cells or stimulated splenocytes. T-cell hybrids are protected from activation-induced apoptosis by a Nur77 dominant negative mutation. Induction of Nur77 mRNA and cell death by apoptosis following treatment of T-cell hybrids with antibody directed against the TCR has also been shown. Additionally, transfection of the T-cell hybrids with antisense Nur77 protects cells from apoptosis when signaled to die by TCR engagement. Nur77 is a phosphoprotein which migrates on SDS/PAGE gels as diffuse bands between 67 and 88kDa depending on post-translational modifications. Clone 12.14 recognizes mouse Nur77. A full-length mouse Nur77 fusion protein was used as immunogen.

Host Species: Mouse
Clone: 12.14
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Full-length mouse Nur77 fusion protein
Formula Weight [Chemical]: 67-88kDa

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nur77
Applications Western Blot
Classification Monoclonal
Clone 12.14
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Nr4a1; NGFIB; NGFI-B; GFRP1; Gfrp; N10; NP10; TR3 ; Hbr1; Hbr-1; Hmr; TIS1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Full-length mouse Nur77 fusion protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.