Learn More
NR4A1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA513296
Description
NR4A1 is a member of the steroid-thyroid hormone-retinoid receptor superfamily. Expression is induced by phytohemagglutinin in human lymphocytes and by serum stimulation of arrested fibroblasts. This protein acts as a nuclear transcription factor. Translocation of the protein from the nucleus to mitochondria induces apoptosis.
This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications
| NR4A1 | |
| Polyclonal | |
| NR4A1 | |
| Orphan nuclear receptor NR4A1, Orphan nuclear receptor HMR, Early response protein NAK1, TR3 orphan receptor, ST-59, NR4A1, GFRP1, HMR, NAK1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3164 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| P22736 | |
| NR4A1 | |
| KLH conjugated synthetic peptide between 329-358 amino acids from human NUR77(NR4A1) | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.