Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NY-ESO-1 Mouse anti-Human, Clone: E978, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05812
Description
Anti-NY-ESO-1 Antibody, clone E978 detects level of NY-ESO-1 and has been published and validated for use in western blotting.
Specifications
NY-ESO-1 | |
Monoclonal | |
Unconjugated | |
NM_001327 | |
Mouse | |
Ascites | |
Primary | |
−20°C for 24 months. | |
IgG |
Western Blot | |
E978 | |
mouse ascites with 0.05% sodium azide | |
CTAG1A, CTAG, CTAG1, ESO1, LAGE2, LAGE2A | |
NY-ESO-1 full-length recombinant protein | |
100 μL | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction