Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
O4 Mouse anti-Chicken, Human, Mouse, Rat, Clone: 81 (also referred to in the literature as mAB O4), MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB345
Description
Anti-O4 Antibody, clone 81 is an antibody against O4 for use in immunocytochemistry, immunohistochemistry with more than 50 product citations.
Specifications
O4 | |
Monoclonal | |
Unconjugated | |
Mouse | |
Ammonium sulfate precipitation | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgM |
Immunocytochemistry, Immunohistochemistry | |
81 (mAB O4) | |
Purified immunoglobulin in 0.05M Potassium phosphate buffer, pH 8.0 with 0.3M NaCl and 0.05% sodium azide. | |
Homogenate of white matter of corpus callosum from bovine brain. | |
50 μg | |
Chicken, Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction