Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OASIS/CREB3L1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131017
Description
OASIS/CREB3L1 Polyclonal specifically detects OASIS/CREB3L1 in Human samples. It is validated for Western Blot.Specifications
OASIS/CREB3L1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
CREB3L1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CREB3L1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
90993 | |
Human | |
IgG |
Western Blot | |
1.0mg/mL | |
Western Blot 1:500-1:3000 | |
BBF-2 homolog, cAMP responsive element binding protein 3-like 1, cAMP-responsive element-binding protein 3-like protein 1, OASIScyclic AMP-responsive element-binding protein 3-like protein 1, old astrocyte specifically induced substance, Old astrocyte specifically-induced substance | |
Rabbit | |
57 kDa | |
100 μL | |
Primary | |
Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (85%) and Rat (85%). Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction