Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OASIS/CREB3L1 Antibody, Novus Biologicals™
SDP

Catalog No. NB600799 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600799 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600799 Supplier Novus Biologicals Supplier No. NB600799
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

OASIS/CREB3L1 Polyclonal specifically detects OASIS/CREB3L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen OASIS/CREB3L1
Applications Western Blot, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_443086.1
Gene Alias BBF-2 homolog, cAMP responsive element binding protein 3-like 1, cAMP-responsive element-binding protein 3-like protein 1, OASIScyclic AMP-responsive element-binding protein 3-like protein 1, old astrocyte specifically induced substance, Old astrocyte specifically-induced substance
Gene Symbols CREB3L1
Host Species Goat
Immunogen Peptide with sequence C-DFSHSKEWFHDRD corresponding to C-Terminus according to NP_443086.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90993
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse and Rat.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.