Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-OCLN Clone: 1G7,

Catalog No. CB1044100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
CB1044100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB1044100UG Supplier MilliporeSigma™ Supplier No. CB1044100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects OCLN Clone: 1G7 in Human samples, and it is validated for ELISA, Immunocytochemistry, Immuno Blotting

OCLN is an integral membrane protein which is located at tight junctions. This protein may be involved in the formation and maintenance of the tight junction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCLN
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry
Classification Monoclonal
Clone 1G7
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen A recombinant polypeptide corresponding to amino acids of 423-523 human OCLN, expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽55-60kDa OCLN protein in HL-60 and HeLa cells.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.