Learn More
occludin, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant OCLN.
Supplier: Abnova Corporation H00004950A01
Description
This gene encodes an integral membrane protein which is located at tight junctions. This protein may be involved in the formation and maintenance of the tight junction. The possibility of several alternatively spliced products has been suggested but the full nature of these products has not been described. [provided by RefSeq
Sequence: ITSDQQRQLYKRNFDTGLQEYKSLQSELDEINKELSRLDKELDDYREESEEYMAAADEYNRLKQVKGSADYKSKKNHCKQLKSKLSHIKKMVGDYDRQKTSpecifications
occludin | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant OCLN. | |
OCLN | |
OCLN | |
OCLN (NP_002529, 423 a.a. ∼ 522 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_002538 | |
Mouse | |
50 μL | |
Yes | |
4950 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.