Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Oct-1, Clone: YL15, R&D Systems™

Catalog No. K01913
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K01913 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. K01913 Supplier R&D Systems Supplier No. K01913
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Applications:

Immunoprecipitation, Western Blotting

Specifications

Antigen OCT1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone YL15
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation
Formulation PBS containing 50% glycerol, pH 7.2 with No Preservative
Gene Alias FLJ42836, NF-A1, Oct-1, OCT1class 2, transcription factor 1, OTF1, POU class 2 homeobox 1, POU domain class 2, transcription factor 1, POU2F1
Gene Symbols POU2F1
Host Species Mouse
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5451
Target Species Human, Mouse
Content And Storage Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.