Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oct-2 Rat, Unlabeled, Clone: 9A2, BD

Catalog No. BDB562837 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562837 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562837 Supplier BD Biosciences Supplier No. 562837

Rat Monoclonal Antibody

The 9A2 monoclonal antibody specifically binds to Oct-2 that has a molecular weight of approximately 50-60 kDa. The 9A2 antibody was generated against the 44 N-terminal amino acids common to mouse Oct-2 isoforms and crossreacts with both human and mouse Oct-2. Oct-2 is a POU class 2 homeobox 2 transcription factors (POU2F2) that is also known as OCT2, OTF2, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2 and Octamer-binding transcription factor 2. The POU domain in the Oct-2 protein is required for low and high affinity cooperative binding of the octomer sequence and heptamer site in immunoglobulin promoters and promoters for other genes. Oct-2 is expressed in B cells, activated CD4+ and CD8+ T cells, monocytes/macrophages and subsets of dendritic cells. Oct-2 is reportedly associated with neoplasms including Diffuse Large B-cell lymphoma and Nodular Lymphocyte predominant Hodgkin's lymphoma. Studies with knockout mouse models suggest Oct-2 is required for B cell maturation, but not for B cell commitment, and for B cell production of IL-6.

Host Species: Rat
Clone: 9A2
Isotype: IgG2a, κ
Species Reactivity: Hu, Ms
Immunogen: Mouse Oct-2 amino acids 1-44 Recombinant Protein

Flow Cytometry, Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen OCT2
Applications Flow Cytometry
Classification Monoclonal
Clone 9A2
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Oct2, OTF-2, OCT2, OTF2, Oct2, NF-A2, POU2F2, Pou2f2, Otf2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Oct-2 amino acids 1-44 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.