Learn More
Oct-2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA138329
Description
Oct-2 is a transcription factor belonging to the POU homeo-domain family that binds to the Ig gene octomer sites regulating B cell specific genes. Oct-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. Oct-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.
This antibody is predicted to react with canine, mouse, porcine, rabbit based on sequence homology.Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
Specifications
Oct-2 | |
Polyclonal | |
Assay Dependent | |
P09086 | |
POU2F2 | |
Synthetic peptide derived from C-terminus of human Oct-2 | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
POU2F2 | |
OCT2, OTF2, Oct-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5452 | |
Store at 4°C, do not freeze | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.