Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oct-4 Mouse anti-Human, AlexaFluor™488, Clone: 10H11.2 , MilliporeSigma™

Catalog No. FCMAB113A4 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB113A4 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB113A4 Supplier MilliporeSigma Supplier No. FCMAB113A4
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Oct-4 using this Anti-Oct-4 Antibody, clone 10H11.2, Alexa Fluor 488 conjugate validated for use in FC and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oct-4.
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 10H11.2
Conjugate Alexa Fluor 488
Formulation Purified mouse monoclonal IgG1 conjugated to Alexa Flour™ 488 in PBS with less than 0.09% sodium azide and 15mg/mL BSA.
Gene Accession No. NM_002701.4
Gene Symbols MGC22487, OCT3, OCT4, OTF3, OTF4, OTTHUMP00000029292, Oct-3, Oct-4 , Oct4
Host Species Mouse
Immunogen Recombinant GST fusion protein, human Oct-4 (aa 1-134).
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.