Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT-4 [POU5F1] Mouse anti-Human, Mouse, AlexaFluor™488, Clone: 7F9.2, MilliporeSigma™

Catalog No. MAB4419A4MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4419A4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4419A4MI Supplier MilliporeSigma Supplier No. MAB4419A4
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-OCT-4 [POU5F1] Antibody, clone 7F9.2, Alexa Fluor 488 conjugate is an antibody against OCT-4 [POU5F1] for use in immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCT-4 [POU5F1]
Applications Immunocytochemistry
Classification Monoclonal
Clone 7F9.2
Conjugate Alexa Fluor 488
Formulation Purified mouse monoclonal IgG1 conjugated to Alexa Fluor™ 488 in PBS with 0.1% sodium azide and 15mg/ml BSA.
Gene Accession No. NP_038661
Gene Symbols OCT3, OCT4, OTF3, OTF4, Oct-3, Oct-4
Host Species Mouse
Immunogen Recombinant human Oct-4 protein, aa 1-145
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.