Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oct-4 Mouse anti-Human, Mouse, Clone: KT110, MilliporeSigma™

Catalog No. MABN738MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN738MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN738MI Supplier MilliporeSigma Supplier No. MABN738
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-OCT6 Antibody, clone KT110 is a highly specific mouse monoclonal antibody, that targets Oct-6 and has been tested in western blotting, immunohistochemistry and Electrophoretic Mobility Shift Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oct-6.
Applications Electromobility Shift Assay, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone KT110
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS 0.1% azide.
Gene Accession No. NP_035271
Gene Symbols Pou3f1, Oct6, Otf-6, Otf6, Scip
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse OCT6.
Purification Method Protein A purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.