Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR16/OR23, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA132856
Description
OR16/OR23 belongs to the Olfactory Receptor subfamily of GPCRs.
PA1-32856 detects Olfactory receptor OR16/OR23 from human samples.PA1-32856 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-32856 immunogen is: Synthetic peptide, KLH conjugated.
Specifications
Olfactory Receptor OR16/OR23 | |
Polyclonal | |
Unconjugated | |
Synthetic peptide, KLH conjugated. | |
25 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Immunohistochemistry (Paraffin) | |
1mg/mL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction