Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olig2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB15328MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15328MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15328MI Supplier MilliporeSigma Supplier No. AB15328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Olig2 Antibody is validated for use in western blotting for the detection of Olig2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olig2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS containing 0.1% sodium azide.
Gene Symbols OLIG2; bHLHB1; RACK17; OLIGO2; BHLHB1; PRKCBP2; Oligo2
Host Species Rabbit
Immunogen Synthetic peptide from rat.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005806.2
Target Species Rat
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.