Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olig2 Mouse anti-Mouse, Human, Rat, Clone: 211F1.1, MilliporeSigma™

Catalog No. MABN50MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN50MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN50MI Supplier MilliporeSigma Supplier No. MABN50
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Olig2 using this Anti-Olig2 Antibody, clone 211F1.1 validated for use in western blot, immunocytochemistry, immunohistochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olig2
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 211F1.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_005797
Gene Symbols OLIG2, OLIGO2, RACK17, PRKCBP2, Oligo2
Host Species Mouse
Immunogen Recombinant protein corresponding to human Olig2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.