Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oligodendrocytes Mouse anti-Chicken, Human, Mouse, Monkey, Rat, Feline, Clone: CE-1, MilliporeSigma™

Catalog No. MAB328MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB328MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB328MI Supplier MilliporeSigma Supplier No. MAB328

Mouse Monoclonal Antibody

This Anti-Oligodendrocytes Antibody, clone CE-1 is validated for use in immunoprecipitation, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) for the detection of Oligodendrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oligodendrocytes
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone CE-1
Conjugate Unconjugated
Host Species Mouse
Immunogen Rat glial membranes and whole brain white matter.
Quantity 100 μL
Primary or Secondary Primary
Target Species Chicken, Feline, Human, Monkey, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.