Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

oxidized low density lipoprotein (lectin-like) receptor 1, Mouse, Clone: 2C9, Abnova™

Catalog No. 89014158 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-158 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-158 Supplier Abnova Corporation Supplier No. H00004973M08
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant OLR1.

This gene encodes a receptor protein which belongs to the C-type lectin superfamily. This gene is regulated through the cyclic AMP signaling pathway. The encoded protein binds, internalizes and degrades oxidized low-density lipoprotein. This protein may be involved in the regulation of Fas-induced apoptosis. This protein may play a role as a scavenger receptor. Mutations of this gene have been associated with atherosclerosis, risk of myocardial infarction, and may modify the risk of Alzheimer's disease. Alternative transcript variants have been described but their full-length sequence has not been determined. [provided by RefSeq]

Sequence: MTFDDLKIQTVKDQPDEKSNGKKAKGLQFLYSPWWCLAAATLGVLCLGLVVTIMVLGMQLSQVSDLLTQEQANLTHQKKKLEGQISARQQAEEASQESENELKEMIETLARKLNEKSKEQMELHHQNLNLQETLKRVANCSAPCPQDWIWHGENCYLFSSGSFNWEKSQEKCLSLDAKLLKINSTADLDFIQQAISYSSFPFWMGLSRRNPSYPWLWEDGSPLMPHLFRVRGAVSQTYPSGTCAYIQRGAVYAENCILAAFSICQKKANLRAQ

Specifications

Antigen oxidized low density lipoprotein (lectin-like) receptor 1
Applications ELISA
Classification Monoclonal
Clone 2C9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant OLR1.
Formulation PBS with no preservative; pH 7.4
Gene OLR1
Gene Accession No. BC022295
Gene Alias CLEC8A/LOX1/SCARE1
Gene Symbols OLR1
Host Species Mouse
Immunogen OLR1 (AAH22295, 1 a.a. ∼ 273 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4973
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.