Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPA1 Mouse, Unlabeled, Clone: 18, BD

Catalog No. BDB612606 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612606 50 μg
BDB612607 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612606 Supplier BD Biosciences Supplier No. 612606
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Three major GTP-binding protein families include trimeric and low molecular weight G-proteins, as well as a family of large proteins homologous to dynamin. The dynamin family contains proteins with diverse structure and function, but highly homologous N-terminal GTPase domains. A subgroup of the dynamin G-protein-binding family includes the mitochondrial proteins Drp1/Dnm1, Mgm1, and OPA1. The latter protein is mutated in dominant optic atrophy, a disease that involves loss of visual acuity and atrophy of the optic nerve. OPA1 is expressed in heart, brain, liver, and kidney. The sequence of OPA1 includes an N-terminal region that contains a mitochondrial targeting domain and three GTP-binding motifs. The overexpression of OPA1 in Cos-7 cells shows co-localization with cytochrome c in mitochondria, and leads to alterations in mitochondrial morphology from a characteristic tubuluar shape to a vesicular pattern. Thus, OPA1 may have roles in mitochondrial biogenesis that are critical for normal cell function. This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen OPA1
Applications Western Blot
Classification Monoclonal
Clone 18
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human OPA1 aa. 708-830
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.