Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Opioid Receptor Rabbit anti-Human, Mouse, Rat, Primate, Polyclonal, MilliporeSigma™

Catalog No. AB5511MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5511MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5511MI Supplier MilliporeSigma Supplier No. AB5511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Opioid Receptor using this Anti-Opioid Receptor Antibody, μ, pain validated for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opioid Receptor
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum in buffer containing 0.05% sodium azide.
Gene Symbols OPRM1; MOR; MOR-1; MOR1; OPRM; KIAA0403
Host Species Rabbit
Immunogen Synthetic peptide from the C-terminus of rat Mu Opioid receptor.
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000914.2
Target Species Human, Mouse, Primate, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.