Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Opioid Receptor μ Guinea Pig anti-Mouse, Rat, Primate, Polyclonal, MilliporeSigma™

Catalog No. AB5509MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5509MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5509MI Supplier MilliporeSigma Supplier No. AB5509
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Anti-Opioid Receptor μ Antibody-AB5509 is an antibody against Opioid Receptor for use in immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opioid Receptor μ
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Guinea pig serum. Liquid containing 0.05% sodium azide.
Gene Symbols OPRM1; MOR-1; MOR1; OPRM; KIAA0403
Host Species Guinea Pig
Immunogen Synthetic peptide (Catalog number AG375) from the C-terminus of rat mu Opioid receptor corresponding to residues 384-398 of the rat mu opioid receptor.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000914.2; NM_001008503.1; NM_001008504.1; NM_001008505.1
Target Species Mouse, Primate, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.