Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Opsin Rabbit anti-Human, Mouse, Monkey, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5405
Description
Anti-Opsin Antibody, Red/Green detects level of Opsin and has been published and validated for use in immunohistochemistry (paraffin).
Specifications
Opsin | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS, 0.02M phosphate, 0.25M NaCl, pH 7.6 with 0.1% sodium azide. | |
Rabbit | |
Protein A chromatography | |
Primary | |
Human, Monkey, Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
OPN1LW; CBP; CBBM; RCP | |
Recombinant human red/green opsin. | |
100 μg | |
NM_020061.3 | |
2-8°C in undiluted aliquots for up to 6 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction