Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Opsin Rabbit anti-Human, Mouse, Monkey, Polyclonal, MilliporeSigma™

Catalog No. AB5407MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5407MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5407MI Supplier MilliporeSigma Supplier No. AB5407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Opsin Antibody, blue is validated for use in immunohistochemistry (paraffin) for the detection of Opsin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opsin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulinin PBS {0.02M phosphate, 0.25M NaCl, pH 7.6} with 0.1% sodium azide as a preservative
Gene Symbols OPN1SW; CBT; BOP; BCP
Host Species Rabbit
Immunogen Recombinant human blue opsin.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001708.1
Target Species Human, Monkey, Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.