Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

olfactory receptor, family 2, subfamily C, member 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005888 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-888 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-888 Supplier Abnova Corporation Supplier No. H00004993B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human OR2C1 protein.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq

Sequence: MDGVNDSSLQGFVLMGISDHPQLEMIFFIAILFSYLLTLLGNSTIILLSRLEARLHTPMYFFLSNLSSLDLAFATSSVPQMLINLWGPGKTISYGGCITQLYVFLWLGATECILLVVMAFDRYVAVCRPLRYTAIMNPQLCWLLAVIAWLGGLGNSVIQSTFTLQLPLCGHRRVEGFLCEVPAMIKLACGDTSLNQAVLNGVCTFFTAVPLSIIVISYCLIAQAVLKIRSAEGRRKAFNTCLSHLLVVFLFYGSASYGYLLPAKNSKQDQGKFISLFYSLVTPMVNPLIYTLRNMEVKGALRRLLGKGREVG

Specifications

Antigen olfactory receptor, family 2, subfamily C, member 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human OR2C1 protein.
Formulation No additive
Gene OR2C1
Gene Accession No. NM_012368.1
Gene Alias MGC163200/MGC95444/OLFmf3/OR2C2P
Gene Symbols OR2C1
Host Species Mouse
Immunogen OR2C1 (AAI30329.1, 1 a.a. ∼ 312 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Membrane Receptors
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4993
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.