Learn More
OR56B1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human OR56B1 peptide using ARM Technology.
Supplier: Abnova Corporation H00387748K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
OR56B1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
OR56B1 | |
OR56B1 | |
A synthetic peptide of human OR56B1 | |
RUO | |
387748 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
olfactory receptor, family 56, subfamily B, member 1 | |
OR56B1 | |
OR11-65, OR56B1P | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.