Learn More
OR6Y1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human OR6Y1 peptide using ARM Technology.
Supplier:  Abnova Corporation H00391112K
Catalog No. 89-204-567
Request a Quote
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line
Order Info
  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
Specifications
OR6Y1
Monoclonal
Unconjugated
In 1X PBS, pH 7.4
OR6Y1
OR6Y1
A synthetic peptide of human OR6Y1
RUO
391112
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG
ELISA, Western Blot
1mg
olfactory receptor, family 6, subfamily Y, member 1
OR6Y1
OR1-11, OR6Y2
Rabbit
1 mg x up to 3 Abs
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.