Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orai1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB9868MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9868MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9868MI Supplier MilliporeSigma Supplier No. AB9868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Orai1 Antibody, Intracellular is validated for use in western blotting for the detection of Orai1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Orai1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin in PBS with 0.1% NaN3
Gene Symbols ORAI1; CRACM1; TMEM142A
Host Species Rabbit
Immunogen Synthetic Peptide from the Intracellular domain of Orai1
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.