Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ORAI3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520375
Description
A suggested positive control is A-20 cell lysate. PA5-20375 can be used with blocking peptide PEP-0492.
ORAI3 is a protein coding gene that is involved in calcium signaling and regulation of calcium homeostasis. It is a member of the ORAI family of proteins that form calcium release-activated calcium (CRAC) channels in the plasma membrane of cells. These channels are activated by depletion of calcium from the endoplasmic reticulum, leading to an influx of calcium ions into the cytoplasm. ORAI3 has been implicated in a variety of physiological processes, including T-cell activation, muscle contraction, and neurotransmitter release. Dysregulation of ORAI3 expression or function has been associated with various diseases, including immune disorders, cancer, and cardiovascular disease.
Specifications
ORAI3 | |
Polyclonal | |
Unconjugated | |
ORAI3 | |
9930124N15; BC061259; calcium release-activated calcium channel 3; calcium release-activated calcium modulator 3 protein; ORAI calcium release-activated calcium modulator 3; ORAI3; Protein orai-3; RGD1306538; Tmem142c; Transmembrane protein 142C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269999, 93129 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6P8G8, Q9BRQ5 | |
ORAI3 | |
A 19 amino acid peptide from near the carboxy terminus of human ORAI3. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction