Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORC6 Rat anti-Human, Clone: 3A4, MilliporeSigma™

Catalog No. 05938MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-938-MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-938-MI Supplier MilliporeSigma Supplier No. 05938
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-ORC6 Antibody, clone 3A4 is a high quality Rat Monoclonal Antibody for the detection of ORC6 and has been validated in Chimmunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC6
Applications ChIP Assay, Western Blot
Classification Monoclonal
Clone 3A4
Conjugate Unconjugated
Gene Accession No. NP_055136
Gene Symbols ORC6L, ORC6
Host Species Rat
Immunogen Full-length human origin recognition complex 6 (ORC6). Clone 3A4.
Purification Method Protein G purfied
Quantity 50 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.