Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Orexin-2 Receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3094
Description
Detect Orexin-2 Receptor using this Anti-Orexin-2 Receptor Antibody validated for use in ELISA, immunohistochemistry and western blotting.
Specifications
| Orexin-2 Receptor | |
| Polyclonal | |
| Affinity Purified 1mg/mL solution in PBS, pH 7.4 and 0.1% BSA | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| HCRTR2; OTTHUMP00000039969; Ox2r; OX2R | |
| A 19 aa peptide near the C-terminus (cytoplasmic domain) of rat OX2R. The immunogen peptide (Catalog Number AG794) is available to perform preabsorbtion controls. | |
| 50 μg | |
| NP_001517 | |
| −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction