Learn More
Orexin-A Rabbit anti-Human, Mouse, Rat, Feline, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Orexin-A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Liquid in PBS, pH 7.4 with 0.1% BSA and 0.1% sodium azide. |
| Gene Symbols | HCRT; Hypocretin; OX; orexin; Hcrt; PPOX; PPORX |
| Host Species | Rabbit |
| Immunogen | An 18 amino acid peptide mapping near the amino terminus of mouse Orexin-A. The immunogen sequence is identical in human, rat, porcine, ovine, bovine and canine and has 81% homology with chicken. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.