Learn More
Orexin-B Rabbit anti-Bovine, Canine, Mouse, Pig, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Orexin-B |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS, with 0.1% BSA . |
| Gene Symbols | HCRT; Hypocretin; OX; orexin; Hcrt; PPOX; PPORX |
| Host Species | Rabbit |
| Immunogen | A 13 amino acid peptide mapping near the amino terminus of mouse Orexin-B. The immunogen peptide has 92% similarity with human Orexin-B. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.