Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organic Anion Transporting Polypeptide 1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3570PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3570PMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3570PMI Supplier MilliporeSigma Supplier No. AB3570P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Organic Anion Transporting Polypeptide 1 Antibody is validated for use in ELISA, western blotting for the detection of Organic Anion Transporting Polypeptide 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Organic Anion Transporting Polypeptide 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA.
Gene Symbols SLCO4A1; OATP1; POAT; OATPE; OATPRP1; OATP-RP1; SLC21A12; OATP4A1; OATP-E
Host Species Rabbit
Immunogen A 17 amino acid peptide sequence near amino acid 285 of rat OATP1 (Jacquemin et al. 1994; Kullak-Ublick et al. 1995).
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_016354.3
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.