Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Organic Anion Transporting Polypeptide 2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3572P
Description
Detect Organic Anion Transporting Polypeptide 2 using this Anti-Organic Anion Transporting Polypeptide 2 Antibody, C-terminus, intracellular validated for use in ELISA and western blotting.
Specifications
Organic Anion Transporting Polypeptide 2 | |
Polyclonal | |
Affinity purified immunoglobulin. Liquid in PBS, containing 0.1% BSA. | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat |
ELISA, Western Blot | |
Unconjugated | |
SLCO1B1; LST1; LST-1; OATP2; OATPC; OATP1B1; SLC21A6; MGC133282; OATP-C | |
A 12 amino acid peptide sequence near the cytoplasmic, C-terminus of rat OATP2. The immunogen shows no significant sequence similarity to other OATPs or other proteins. | |
50 μg | |
NM_006446.3 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction