Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

oncostatin M receptor, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018643 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-643 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-643 Supplier Abnova Corporation Supplier No. H00009180D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human OSMR protein.

Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF [provided by RefSeq

Sequence: MALFAVFQTTFFLTLLSLRTYQSEVLAERLPLTPVSLKVSTNSTRQSLHLQWTVHNLPYHQELKMVFQIQISRIETSNVIWVGNYSTTVKWNQVLHWSWESELPLECATHFVRIKSLVDDAKFPEPNFWSNWSSWEEVSVQDSTGQDILFVFPKDKLVEEGTNVTICYVSRNIQNNVSCYLEGKQIHGEQLDPHVTAFNLNSVPFIRNKGTNIYCEASQGNVSEGMKGIVLFVSKVLEEPKDFSCETEDFKTLHCTWDPGTDTALGWSKQPSQSYTLFESFSGEKKLCTHKNWCNWQITQDSQETYNFTLIAENYLRKRSVNILFNLTHRGETRVVTAHRGH

Specifications

Antigen oncostatin M receptor
Applications In situ PLA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human OSMR protein.
Formulation PBS with no preservative; pH 7.4
Gene OSMR
Gene Accession No. BC010943
Gene Alias MGC150626/MGC150627/MGC75127/OSMRB
Gene Symbols OSMR
Host Species Rabbit
Immunogen OSMR (AAH10943.1, 1 a.a. ∼ 342 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Receptors
Primary or Secondary Primary
Gene ID (Entrez) 9180
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.