Learn More
Osteopontin Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1870
Description
This Anti-Osteopontin Antibody is validated for use in ELISA, immunohistochemistry, immunohistochemistry (paraffin), western blotting for the detection of Osteopontin.
Specifications
Osteopontin | |
Polyclonal | |
Rabbit polyclonal antiserum in buffer containing no preservatives. | |
Rabbit | |
Unpurified | |
Primary | |
Human, Mouse, Rat | |
Serum |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SPP1; BNSP; MGC110940; Uropontin; Nephropontin; CIT:779436; osteopontin; SPP-1; BSPI; OPN; ETA-1 | |
Synthetic peptide corresponding to amino acids 75-90 of human osteopontin (SwissProt Accession # P10451; PSKSNESHDHMDDM DD). | |
100 μL | |
NP_000573 | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |