Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteopontin Rat anti-Human, Clone: 4AA, MilliporeSigma™

Catalog No. MAB3055MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3055MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3055MI Supplier MilliporeSigma Supplier No. MAB3055

Rat Monoclonal Antibody

Anti-Osteopontin Antibody, recombinant protein only, clone 4AA is an antibody against Osteopontin for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Osteopontin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4AA
Conjugate Unconjugated
Formulation Purified immunoglobulin in PBS containing sodium azide.
Gene Accession No. NM_000582.2; NM_001040058.1; NM_001040060.1
Gene Symbols SPP1, BNSP, osteopontin, SPP-1, BSPI, OPN, ETA-1, MGC110940, Uropontin, Nephropontin, CIT:779436
Host Species Rat
Immunogen Antibody MAB3055 has been generated against recombinant human osteopontin (Hu, 1995)
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.