Learn More
MilliporeSigma™ anti-Osteopontin (OPN; BSP-1; SPP1; Uropontin); Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB1870/DEL
Description
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting
Specifications
Osteopontin/OPN/BSP-1 | |
Unconjugated | |
Synthetic peptide corresponding to amino acids 75-90 of human osteopontin (SwissProt Accession P10451; PSKSNESHDHMDDMDD). | |
RUO | |
Primary | |
Serum |
Polyclonal | |
Rabbit | |
100 μL | |
Cell Structure | |
Human, Mouse, Rat |