Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

orthodenticle homeobox 1, Mouse, Clone: 4E5, Abnova™

Catalog No. 89000852 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-852 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-852 Supplier Abnova Corporation Supplier No. H00005013M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant OTX1.

This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. A similar protein in mice is required for proper brain and sensory organ development and can cause epilepsy. [provided by RefSeq

Sequence: YGMNGLGLAGPAMDLLHPSVGYPATPRKQRRERTTFTRSQLDVLEALFAKTRYPDIFMREEVALKINLPESRVQVWFKNRRAKCRQQQQSGSGTKSRPAKKKSSPVR

Specifications

Antigen orthodenticle homeobox 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant OTX1.
Formulation PBS with no preservative; pH 7.4
Gene OTX1
Gene Accession No. NM_014562
Gene Alias FLJ38361/MGC15736
Gene Symbols OTX1
Host Species Mouse
Immunogen OTX1 (NP_055377, 10 a.a. ∼ 116 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5013
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.