Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-OV-6, PE, Clone: OV-6, R&D Systems™
Supplier: R&D Systems FAB2020P
This item is not returnable.
View return policy
Description
Applications:
Flow Cytometry
Specifications
OV-6 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Carcinogen-treated rat nodular hepatocytes | |
100 Tests | |
Primary | |
Human, Rat | |
Purified |
Flow Cytometry | |
OV-6 | |
Flow Cytometry 10 uL/10^6 cells | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects human and rat OV-6 antigen. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction